Merck
  • Influence of susceptibility to hydrolysis and hydrophobicity of arylsemicarbazones on their anti-nociceptive and anti-inflammatory activities.
Influence of susceptibility to hydrolysis and hydrophobicity of arylsemicarbazones on their anti-nociceptive and anti-inflammatory activities.
European journal of medicinal chemistry (2012-02-24)
Rafael P Vieira, Josane A Lessa, Wallace C Ferreira, Fernanda B Costa, Leandro F S Bastos, Willian R Rocha, Márcio M Coelho, Heloisa Beraldo
ZUSAMMENFASSUNG
Benzaldehyde semicarbazone (BS) inhibited zymosan writhing response, carrageenan paw edema and both phases of formaldehyde nociceptive response. 2-hydroxybenzaldehyde semicarbazone (2-OHBS) and semicarbazide inhibited carrageenan paw edema and the second phase of formaldehyde nociceptive response. 2-OHBS inhibited zymosan writhing response. 3- and 4-OHBS did not show such activities. 2-OHBS showed the lowest LUMO energy, the highest contribution of the iminic carbon to LUMO energy, the highest positive charge on the iminic carbon, the highest negative charge on the iminic nitrogen and the highest susceptibility to hydrolysis. Hence semicarbazide may play important roles in 2-OHBS's activities. Inhibition of the first phase of formaldehyde response by BS could be attributed to its higher hydrophobicity and lower susceptibility to hydrolysis in comparison to 2-OHBS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzaldehyd, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyd, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyd, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyd, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyd, analytical standard
Sigma-Aldrich
Benzaldehyd, natural, FCC, FG