Merck
  • Antimicrobial activity of sulfonamides containing 5-chloro-2-hydroxybenzaldehyde and 5-chloro-2-hydroxybenzoic acid scaffold.
Antimicrobial activity of sulfonamides containing 5-chloro-2-hydroxybenzaldehyde and 5-chloro-2-hydroxybenzoic acid scaffold.
European journal of medicinal chemistry (2012-03-01)
Martin Krátký, Jarmila Vinšová, Marie Volková, Vladimír Buchta, František Trejtnar, Jiřina Stolaříková
ZUSAMMENFASSUNG
A series of novel sulfonamides containing 5-chloro-2-hydroxybenzaldehyde or 5-chloro-2-hydroxybenzoic acid scaffolds were designed, synthesized and characterized by IR, (1)H NMR and (13)C NMR. All ten target synthesized derivatives and starting sulfonamides were evaluated in vitro for the activity against Gram-positive and Gram-negative bacteria, fungi, Mycobacterium tuberculosis, Mycobacterium avium and Mycobacterium kansasii. The most active compound against methicillin-sensitive and methicillin-resistant Staphyloccoccus aureus was 5-chloro-N-{4-[N-(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl}-2-hydroxybenzamide with MIC 15.62-31.25 μmol/L. 4-Amino-N-(thiazol-2-yl)benzenesulfonamide and 4-(5-chloro-2-hydroxybenzylideneamino)-N-(thiazol-2-yl)benzenesulfonamide have shown the best activity against M. kansasii at the concentrations of 1-4 μmol/L. The efficacy against other strains was weaker and the studied derivatives exhibited almost none antifungal potency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Sulfamethoxazol
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Sulfamethazin, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Sulfadiazin, 99.0-101.0%
Supelco
Sulfamethoxazol, VETRANAL®, analytical standard
Supelco
Sulfamethazin, VETRANAL®, analytical standard
Sigma-Aldrich
Bacitracin Zinksalz, from Bacillus licheniformis, ~70,000 U/g
Supelco
Sulfathiazol
Supelco
Sulfadiazin, VETRANAL®, analytical standard
Supelco
Sulfathiazol, VETRANAL®, analytical standard