Merck
  • Comparative epigenetic analysis of tumour initiating cells and syngeneic EPSC-derived neural stem cells in glioblastoma.
Comparative epigenetic analysis of tumour initiating cells and syngeneic EPSC-derived neural stem cells in glioblastoma.
Nature communications (2021-10-23)
Claire Vinel, Gabriel Rosser, Loredana Guglielmi, Myrianni Constantinou, Nicola Pomella, Xinyu Zhang, James R Boot, Tania A Jones, Thomas O Millner, Anaelle A Dumas, Vardhman Rakyan, Jeremy Rees, Jamie L Thompson, Juho Vuononvirta, Suchita Nadkarni, Tedani El Assan, Natasha Aley, Yung-Yao Lin, Pentao Liu, Sven Nelander, Denise Sheer, Catherine L R Merry, Federica Marelli-Berg, Sebastian Brandner, Silvia Marino
ZUSAMMENFASSUNG
Epigenetic mechanisms which play an essential role in normal developmental processes, such as self-renewal and fate specification of neural stem cells (NSC) are also responsible for some of the changes in the glioblastoma (GBM) genome. Here we develop a strategy to compare the epigenetic and transcriptional make-up of primary GBM cells (GIC) with patient-matched expanded potential stem cell (EPSC)-derived NSC (iNSC). Using a comparative analysis of the transcriptome of syngeneic GIC/iNSC pairs, we identify a glycosaminoglycan (GAG)-mediated mechanism of recruitment of regulatory T cells (Tregs) in GBM. Integrated analysis of the transcriptome and DNA methylome of GBM cells identifies druggable target genes and patient-specific prediction of drug response in primary GIC cultures, which is validated in 3D and in vivo models. Taken together, we provide a proof of principle that this experimental pipeline has the potential to identify patient-specific disease mechanisms and druggable targets in GBM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Insulin -Lösung human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Millipore
Accutase Zellablösungslösung, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Sigma-Aldrich
Accumax -Lösung, sterile-filtered, suitable for cell culture
Sigma-Aldrich
N6,2′-O-Dibutyryladenosin 3′,5′-zyklisches Monophosphat Natriumsalz Monohydrat, ≥97% (HPLC), powder
Sigma-Aldrich
3,3′,5-Triiod-L-thyronin, ≥95% (HPLC), powder
Sigma-Aldrich
Smoothened Agonist, SAG, A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).