Merck
  • Human exhaled air can efficiently support in vitro maturation of porcine oocytes and subsequent early embryonic development.
Human exhaled air can efficiently support in vitro maturation of porcine oocytes and subsequent early embryonic development.
Animal reproduction (2018-08-16)
Zubing Cao, Di Gao, Tengteng Xu, Xu Tong, Yiqing Wang, Yunsheng Li, Fugui Fang, Jianping Ding, Xiaorong Zhang, Yunhai Zhang
ZUSAMMENFASSUNG
Air phase is an indispensable environmental factor affecting oocyte maturation and early embryo development. Human exhaled air was previously proved to be a reliable and inexpensive atmosphere that sustains normal early development of mouse and bovine embryos. However, whether human exhaled air can support in vitro maturation (IVM) of porcine oocytes is not yet known. To evaluate the feasibility of maturing oocytes in human exhaled air, we examined oocyte morphology, BMP15 expression, nuclear and cytoplasmic maturation. We found that cumulus expansion status, expression levels of BMP15 important for cumulus expansion and the rate of first polar body emission were similar among human exhaled air, 5% O2 or 20% O2 in air after IVM of 44 h. Furthermore, the percentage of metaphase II (MII) oocytes showing normal cortical and sub-membranous localization of cortical granules and diffused mitochondrial distribution patterns is comparable among groups. The cleavage, blastocyst rate and total cell number were not apparently different for parthenogenetic activated and somatic cloned embryos derived from MII oocytes matured in three air phases, suggesting oocytes matured in human exhaled air obtain normal developmental competence. Taken together, human exhaled air can efficiently support in vitro maturation of porcine oocytes and subsequent early embryonic development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lektin aus Arachis hypogaea (Erdnuss), FITC conjugate, lyophilized powder