Merck
The 14-3-3ζ-c-Src-integrin-β3 complex is vital for platelet activation.
Blood (2020-06-26)
Chuanbin Shen, Ming Liu, Runjia Xu, Gan Wang, June Li, Pingguo Chen, Wenjing Ma, James Mwangi, Qiumin Lu, Zilei Duan, Zhiye Zhang, Fatima Zohra Dahmani, Daniel Thomas Mackeigan, Heyu Ni, Ren Lai
ZUSAMMENFASSUNG
Several adaptor molecules bind to cytoplasmic tails of β-integrins and facilitate bidirectional signaling, which is critical in thrombosis and hemostasis. Interfering with integrin-adaptor interactions spatially or temporally to inhibit thrombosis without affecting hemostasis is an attractive strategy for the development of safe antithrombotic drugs. We show for the first time that the 14-3-3ζ-c-Src-integrin-β3 complex is formed during platelet activation. 14-3-3ζ-c-Src interaction is mediated by the -PIRLGLALNFSVFYYE- fragment (PE16) on the 14-3-3ζ and SH2-domain on c-Src, whereas the 14-3-3ζ-integrin-β3 interaction is mediated by the -ESKVFYLKMKGDYYRYL- fragment (EL17) on the 14-3-3ζ and -KEATSTF- fragment (KF7) on the β3-integrin cytoplasmic tail. The EL17-motif inhibitor, or KF7 peptide, interferes with the formation of the 14-3-3ζ-c-Src-integrin-β3 complex and selectively inhibits β3 outside-in signaling without affecting the integrin-fibrinogen interaction, which suppresses thrombosis without causing significant bleeding. This study characterized a previously unidentified 14-3-3ζ-c-Src-integrin-β3 complex in platelets and provided a novel strategy for the development of safe and effective antithrombotic treatments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
RIPA-Puffer
Sigma-Aldrich
Fibrinogen aus Humanplasma, 50-70% protein (≥80% of protein is clottable)