Merck
  • Long-term exposure to a hypomagnetic field attenuates adult hippocampal neurogenesis and cognition.
Long-term exposure to a hypomagnetic field attenuates adult hippocampal neurogenesis and cognition.
Nature communications (2021-02-21)
Bingfang Zhang, Lei Wang, Aisheng Zhan, Min Wang, Lanxiang Tian, Weixiang Guo, Yongxin Pan
ZUSAMMENFASSUNG
Adult hippocampal neurogenesis contributes to learning and memory, and is sensitive to a variety of environmental stimuli. Exposure to a hypomagnetic field (HMF) influences the cognitive processes of various animals, from insects to human beings. However, whether HMF exposure affect adult hippocampal neurogenesis and hippocampus-dependent cognitions is still an enigma. Here, we showed that male C57BL/6 J mice exposed to HMF by means of near elimination of the geomagnetic field (GMF) exhibit significant impairments of adult hippocampal neurogenesis and hippocampus-dependent learning, which is strongly correlated with a reduction in the content of reactive oxygen species (ROS). However, these deficits seen in HMF-exposed mice could be rescued either by elevating ROS levels through pharmacological inhibition of ROS removal or by returning them back to GMF. Therefore, our results suggest that GMF plays an important role in adult hippocampal neurogenesis through maintaining appropriate endogenous ROS levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
5-Brom-2′-deoxyuridin, ≥99% (HPLC)
Sigma-Aldrich
bisBenzimid H 33342, ≥98% (HPLC)
Sigma-Aldrich
Apocynin, A cell-permeable, anti-inflammatory phenolic compound that acts as a potent and selective inhibitor of NADPH oxidase.
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)