Merck
  • Antibacterial activity in secondary metabolite extracts of heterotrophic bacteria against  Vibrio alginolyticus, Aeromonas hydrophila, and  Pseudomonas aeruginosa.
Antibacterial activity in secondary metabolite extracts of heterotrophic bacteria against  Vibrio alginolyticus, Aeromonas hydrophila, and  Pseudomonas aeruginosa.
F1000Research (2021-02-05)
Jarod Setiaji, Feli Feliatra, Hilwan Yuda Teruna, Iesje Lukistyowati, Indra Suharman, Zainal Abidin Muchlisin, Teuku Iskandar Johan
ZUSAMMENFASSUNG
Background: Disease causing bacteria such as Vibrio alginolyticus, Aeromonas hydrophila, and Pseudomonas aeruginosa present a problem for fish farming. Treatment to remove them are generally carried out using antibiotics which have side effects on fish, the environment and humans. However, the use of antibacterial compounds derived from heterotrophic bacteria serve as a good alternative for antibiotics. Therefore, this study aimed to explore antibacterial activity in the secondary metabolite extracts of heterotrophic bacteria against Vibrio alginolyticus, Aeromonas hydrophila, and Pseudomonas aeruginosa. Methods: Heterotrophic bacteria namely Bacillus sp. JS04 MT102913.1, Bacillus toyonensis JS08 MT102920.1, Bacillus cereus JS10 MT102922.1, Bacillus sp. JS11 MT102923.1, Pseudoalteromonas sp. JS19 MT102924.1, Bacillus cereus JS22 MT102926.1, and Bacillus sp. strain JS25 MT102927.1 were used in this study. The sequences of these bacteria have been deposited and are available from NCBI GenBank. Each heterotrophic bacterium was cultured on 6L nutrient broth for 8 days, and extracts produced using ethyl acetate to obtain their secondary metabolites. These extracts were tested for their phytochemical contents using FT-IR and also tested for their inhibitory property in pathogenic bacteria by agar diffusion method. Results: Phytochemical test results showed that the seven heterotrophic bacterial isolates produced terpenoid compounds. Based on the inhibitory test, the secondary metabolite extracts from Bacillus sp strain JS04 had the highest inhibitory effect on the growth of pathogenic bacteria namely, V. alginolyticus (17.5 mm), A. hydrophila (16.8 mm), and P. aeruginosa (17.3 mm). Conclusion: It was concluded that the secondary metabolite extracts of heterotrophic bacteria inhibit the growth of V. alginolyticus, A. hydrophila, and P. aeruginosa.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ReBlot Plus Kit