Merck
Porin 1 Modulates Autophagy in Yeast.
Cells (2021-09-29)
Filomena Broeskamp, Elizabeth S M Edrich, Oskar Knittelfelder, Lisa Neuhaus, Thorsten Meyer, Jonas Heyden, Lukas Habernig, Florian Kreppel, Campbell W Gourlay, Patrick Rockenfeller
ZUSAMMENFASSUNG
Autophagy is a cellular recycling program which efficiently reduces the cellular burden of ageing. Autophagy is characterised by nucleation of isolation membranes, which grow in size and further expand to form autophagosomes, engulfing cellular material to be degraded by fusion with lysosomes (vacuole in yeast). Autophagosomal membranes do not bud from a single cell organelle, but are generated de novo. Several lipid sources for autophagosomal membranes have been identified, but the whole process of their generation is complex and not entirely understood. In this study, we investigated how the mitochondrial outer membrane protein porin 1 (Por1), the yeast orthologue of mammalian voltage-dependent anion channel (VDAC), affects autophagy in yeast. We show that POR1 deficiency reduces the autophagic capacity and leads to changes in vacuole and lipid homeostasis. We further investigated whether limited phosphatidylethanolamine (PE) availability in por1∆ was causative for reduced autophagy by overexpression of the PE-generating phosphatidylserine decarboxylase 1 (Psd1). Altogether, our results show that POR1 deficiency is associated with reduced autophagy, which can be circumvented by additional PSD1 overexpression. This suggests a role for Por1 in Psd1-mediated autophagy regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)