Merck
Competitive binding of E3 ligases TRIM26 and WWP2 controls SOX2 in glioblastoma.
Nature communications (2021-11-05)
Tatenda Mahlokozera, Bhuvic Patel, Hao Chen, Patrick Desouza, Xuan Qu, Diane D Mao, Daniel Hafez, Wei Yang, Rukayat Taiwo, Mounica Paturu, Afshin Salehi, Amit D Gujar, Gavin P Dunn, Nima Mosammaparast, Allegra A Petti, Hiroko Yano, Albert H Kim
ZUSAMMENFASSUNG
The pluripotency transcription factor SOX2 is essential for the maintenance of glioblastoma stem cells (GSC), which are thought to underlie tumor growth, treatment resistance, and recurrence. To understand how SOX2 is regulated in GSCs, we utilized a proteomic approach and identified the E3 ubiquitin ligase TRIM26 as a direct SOX2-interacting protein. Unexpectedly, we found TRIM26 depletion decreased SOX2 protein levels and increased SOX2 polyubiquitination in patient-derived GSCs, suggesting TRIM26 promotes SOX2 protein stability. Accordingly, TRIM26 knockdown disrupted the SOX2 gene network and inhibited both self-renewal capacity as well as in vivo tumorigenicity in multiple GSC lines. Mechanistically, we found TRIM26, via its C-terminal PRYSPRY domain, but independent of its RING domain, stabilizes SOX2 protein by directly inhibiting the interaction of SOX2 with WWP2, which we identify as a bona fide SOX2 E3 ligase in GSCs. Our work identifies E3 ligase competition as a critical mechanism of SOX2 regulation, with functional consequences for GSC identity and maintenance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Hoechst 33258 -Lösung, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
MG-132, ≥95 % durch HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Normal Goat IgG, This Normal Goat IgG is validated for use in ELISA Flow Cytometry Immunoblotting Immunofluorescence Immunohistochemistry Immunoprecipitation for the detection of Goat IgG, Non-immune.