Merck
  • KDM5 Histone Demethylase Activity Links Cellular Transcriptomic Heterogeneity to Therapeutic Resistance.
KDM5 Histone Demethylase Activity Links Cellular Transcriptomic Heterogeneity to Therapeutic Resistance.
Cancer cell (2018-11-26)
Kunihiko Hinohara, Hua-Jun Wu, Sébastien Vigneau, Thomas O McDonald, Kyomi J Igarashi, Kimiyo N Yamamoto, Thomas Madsen, Anne Fassl, Shawn B Egri, Malvina Papanastasiou, Lina Ding, Guillermo Peluffo, Ofir Cohen, Stephen C Kales, Madhu Lal-Nag, Ganesha Rai, David J Maloney, Ajit Jadhav, Anton Simeonov, Nikhil Wagle, Myles Brown, Alexander Meissner, Piotr Sicinski, Jacob D Jaffe, Rinath Jeselsohn, Alexander A Gimelbrant, Franziska Michor, Kornelia Polyak
ZUSAMMENFASSUNG
Members of the KDM5 histone H3 lysine 4 demethylase family are associated with therapeutic resistance, including endocrine resistance in breast cancer, but the underlying mechanism is poorly defined. Here we show that genetic deletion of KDM5A/B or inhibition of KDM5 activity increases sensitivity to anti-estrogens by modulating estrogen receptor (ER) signaling and by decreasing cellular transcriptomic heterogeneity. Higher KDM5B expression levels are associated with higher transcriptomic heterogeneity and poor prognosis in ER+ breast tumors. Single-cell RNA sequencing, cellular barcoding, and mathematical modeling demonstrate that endocrine resistance is due to selection for pre-existing genetically distinct cells, while KDM5 inhibitor resistance is acquired. Our findings highlight the importance of cellular phenotypic heterogeneity in therapeutic resistance and identify KDM5A/B as key regulators of this process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Sigma-Aldrich
Anti-Dimethylhiston-H3-(Lys4)-Antikörper, Upstate®, from rabbit
Millipore
Inosin, A metabolite of adenosine that acts as a potent coronary vasodilator. Exhibits a positive inotropic action on heart muscle and can antagonize the ouabain-induced arrhythmias.
Sigma-Aldrich
Anti-KDM5B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution