Merck
Endocytic BDNF secretion regulated by Vamp3 in astrocytes.
Scientific reports (2021-10-29)
Jeongho Han, Sungryeong Yoon, Hyungju Park
ZUSAMMENFASSUNG
Brain-derived neurotrophic factor (BDNF) regulates diverse brain functions via TrkB receptor signaling. Due to the expression of TrkB receptors, astrocytes can internalize extracellular BDNF proteins via receptor-mediated endocytosis. Endocytosed BDNF can be re-secreted upon stimulation, but the molecular mechanism underlying this phenomenon remains unrecognized. Our study reveals that vesicle-associated membrane protein 3 (Vamp3) selectively regulates the release of endocytic BDNF from astrocytes. By using quantum dot (QD)-conjugated mature BDNF (QD-BDNF) as a proxy for the extracellular BDNF protein, we monitored the uptake, transport, and secretion of BDNF from cultured cortical astrocytes. Our data showed that endocytic QD-BDNF particles were enriched in Vamp3-containing vesicles in astrocytes and that ATP treatment sufficiently triggered either the antero- or retrograde transport and exocytosis of QD-BDNF-containing vesicles. Downregulation of Vamp3 expression disrupted endocytic BDNF secretion from astrocytes but did not affect uptake or transport. Collectively, these results provide evidence of the selective ability of astrocytic Vamp3 to control endocytic BDNF secretion during BDNF recycling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosin-5′-triphosphat Magnesiumsalz, ≥95%, bacterial
Sigma-Aldrich
Ionomycin Calciumsalz, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
Supelco
Polyethylenimin -Lösung, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Heparin-Binding EGF-Like Growth Factor human, HB-EGF, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture