Merck
  • Ester synthesis catalyzed by Mucor miehei lipase immobilized on magnetic polysiloxane-polyvinyl alcohol particles.
Ester synthesis catalyzed by Mucor miehei lipase immobilized on magnetic polysiloxane-polyvinyl alcohol particles.
Applied biochemistry and biotechnology (2004-04-01)
Laura M Bruno, José L de Lima Filho, Eduardo H de M Melo, Heizir F de Castro
ZUSAMMENFASSUNG
Mucor miehei lipase was immobilized on magnetic polysiloxane-polyvinyl alcohol particles by covalent binding with high activity recovered. The performance of the resulting immobilized biocatalyst was evaluated in the synthesis of flavor esters using heptane as solvent. The impact on reaction rate was determined for enzyme concentration, molar ratio of the reactants, carbon chain length of the reactants, and alcohol structure. Ester synthesis was maximized for substrates containing excess acyl donor and lipase loading of 25 mg/mL. The biocatalyst selectivity for the carbon chain length was found to be different concerning the organic acids and alcohols. High reaction rates were achieved for organic acids with 8 or 10 carbons, whereas increasing the alcohol carbon chain length from 4 to 8 carbons gave much lower esterification yields. Optimal reaction rate was determined for the synthesis of butyl caprylate (12 carbons). Esterification performance was also dependent on the alcohol structure, with maximum activity occurring for primary alcohol. Secondary and tertiary alcohols decreased the reaction rates by more than 40%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexansäure-butylester, ≥98%, FG
Sigma-Aldrich
Lipase aus Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)