Merck
Auto-aggressive CXCR6+ CD8 T cells cause liver immune pathology in NASH.
Nature (2021-03-26)
Michael Dudek, Dominik Pfister, Sainitin Donakonda, Pamela Filpe, Annika Schneider, Melanie Laschinger, Daniel Hartmann, Norbert Hüser, Philippa Meiser, Felix Bayerl, Donato Inverso, Jennifer Wigger, Marcial Sebode, Rupert Öllinger, Roland Rad, Silke Hegenbarth, Martina Anton, Adrien Guillot, Andrew Bowman, Danijela Heide, Florian Müller, Pierluigi Ramadori, Valentina Leone, Cristina Garcia-Caceres, Tim Gruber, Gabriel Seifert, Agnieszka M Kabat, Jan-Philipp Mallm, Simon Reider, Maria Effenberger, Susanne Roth, Adrian T Billeter, Beat Müller-Stich, Edward J Pearce, Friedrich Koch-Nolte, Rafael Käser, Herbert Tilg, Robert Thimme, Tobias Boettler, Frank Tacke, Jean-Francois Dufour, Dirk Haller, Peter J Murray, Ron Heeren, Dietmar Zehn, Jan P Böttcher, Mathias Heikenwälder, Percy A Knolle
ZUSAMMENFASSUNG
Nonalcoholic steatohepatitis (NASH) is a manifestation of systemic metabolic disease related to obesity, and causes liver disease and cancer1,2. The accumulation of metabolites leads to cell stress and inflammation in the liver3, but mechanistic understandings of liver damage in NASH are incomplete. Here, using a preclinical mouse model that displays key features of human NASH (hereafter, NASH mice), we found an indispensable role for T cells in liver immunopathology. We detected the hepatic accumulation of CD8 T cells with phenotypes that combined tissue residency (CXCR6) with effector (granzyme) and exhaustion (PD1) characteristics. Liver CXCR6+ CD8 T cells were characterized by low activity of the FOXO1 transcription factor, and were abundant in NASH mice and in patients with NASH. Mechanistically, IL-15 induced FOXO1 downregulation and CXCR6 upregulation, which together rendered liver-resident CXCR6+ CD8 T cells susceptible to metabolic stimuli (including acetate and extracellular ATP) and collectively triggered auto-aggression. CXCR6+ CD8 T cells from the livers of NASH mice or of patients with NASH had similar transcriptional signatures, and showed auto-aggressive killing of cells in an MHC-class-I-independent fashion after signalling through P2X7 purinergic receptors. This killing by auto-aggressive CD8 T cells fundamentally differed from that by antigen-specific cells, which mechanistically distinguishes auto-aggressive and protective T cell immunity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Roche
DNase I, rekombinant, RNase-frei, from bovine pancreas, expressed in Pichia pastoris
Roche
Dispase® II (neutrale Protease, Klasse II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Natriumbutyrat, 98%
Roche
DAPI, 4′,6-Diamidine-2′-phenylindole dihydrochloride
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Probenecid
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Anti-Iba1/AIF1-Antikörper, clone 20A12.1, from mouse
Sigma-Aldrich
U0126, CAS-Nr. 109511-58-2, Calbiochem, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
Wortmannin aus Penicillium funiculosum, from Penicillium funiculosum, ≥98% (HPLC and TLC)