Merck
Formyl peptide receptor 1 signaling potentiates inflammatory brain injury.
Science translational medicine (2021-08-06)
Zhiguo Li, Yulin Li, Jinrui Han, Zilong Zhu, Minshu Li, Qiang Liu, Yongjun Wang, Fu-Dong Shi
ZUSAMMENFASSUNG
Acute brain insults elicit pronounced inflammation that amplifies brain damage in intracerebral hemorrhage (ICH). We profiled perihematomal tissue from patients with ICH, generating a molecular landscape of the injured brain, and identified formyl peptide receptor 1 (FPR1) as the most abundantly increased damage-associated molecular pattern (DAMP) receptor, predominantly expressed by microglia. Circulating mitochondrial N-formyl peptides, endogenous ligands of FPR1, were augmented and correlated with the magnitude of brain edema in patients with ICH. Interactions of formyl peptides with FPR1 activated microglia, boosted neutrophil recruitment, and aggravated neurological deficits in two mouse models of ICH. We created an FPR1 antagonist T-0080 that can penetrate the brain and bind both human and murine FPR1. T-0080 attenuated brain edema and improved neurological outcomes in ICH models. Thus, FPR1 orchestrates brain inflammation after ICH and could be targeted to improve clinical outcome in patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
T-0080, ≥98% (HPLC)