Merck
  • Association Between 17β-Estradiol Receptors and Nitric Oxide Signaling Augments High-Altitude Adaptation of Ladakhi Highlanders.
Association Between 17β-Estradiol Receptors and Nitric Oxide Signaling Augments High-Altitude Adaptation of Ladakhi Highlanders.
High altitude medicine & biology (2021-02-20)
Pooja, Vandana Sharma, Manish Sharma, Rajeev Varshney, Bhuvnesh Kumar, Niroj Kumar Sethy
ZUSAMMENFASSUNG
Pooja, Vandana Sharma, Manish Sharma, Rajeev Varshney, Bhuvnesh Kumar, and Niroj Kumar Sethy. Association between 17β-estradiol receptors and nitric oxide signaling augments high-altitude adaptation of Ladakhi highlanders. High Alt Med Biol. 22: 174-183, 2021. Background: Genomic studies have identified positive natural selection of plasma membrane estrogen receptor signaling pathway for Himalayan highlanders. We sought to investigate significance of this pathway for high-altitude adaptation by studying Ladakhi highlanders. Materials and Methods: We recruited 25 healthy Ladakhi males (age range: 19-37, height: 164 ± 6 cm, and weight 59 ± 4 kg) at Leh (altitude 3,520 m) and age matched sea level volunteers at Delhi (altitude 215 m), India. We evaluated circulatory levels of 17β-estradiol (E2) and testosterone (T) and levels of E2 biosynthesis pathway proteins. In addition, we analyzed mRNA levels of E2 pathway genes and their association with nitric oxide (NO) availability. Results: We observed higher circulatory E2 and lower testosterone (T) in Ladakhi highlanders compared to lowlanders. Studying E2 pathway genes, we identified higher transcript levels of E2 receptors ESR1 (2.02-fold) and ESR2 (3.87-fold) in Ladakhi highlanders. Higher NOS3 mRNA, plasma level of endothelial NO synthase (eNOS), p-eNOS Ser1177, NOx (nitrate and nitrite), and cGMP were observed for Ladakhi highlanders. In addition, we observed a positive correlation between E2 with plasma NOx (r = 0.52, p = 0.002) and cGMP (r = 0.72, p = 0.007) for Ladakhi highlanders. Conclusion: Our results demonstrate higher circulatory E2 and lower T levels in Ladakhi highlanders. Higher levels of E2 and its receptors (ESR1 and ESR2) are positively associated with observed higher levels of eNOS signaling pathway metabolites. These results highlight the functional importance of E2 and its receptors for Himalayan pattern of high-altitude adaptation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid