Merck
  • Gαi/o-coupled Htr2c in the paraventricular nucleus of the hypothalamus antagonizes the anorectic effect of serotonin agents.
Gαi/o-coupled Htr2c in the paraventricular nucleus of the hypothalamus antagonizes the anorectic effect of serotonin agents.
Cell reports (2021-11-18)
Eun-Seon Yoo, Li Li, Lin Jia, Caleb C Lord, Charlotte E Lee, Shari G Birnbaum, Claudia R Vianna, Eric D Berglund, Kathryn A Cunningham, Yong Xu, Jong-Woo Sohn, Chen Liu
ZUSAMMENFASSUNG
The anorexigenic effect of serotonergic compounds has largely been attributed to activation of serotonin 2C receptors (Htr2cs). Using mouse genetic models in which Htr2c can be selectively deleted or restored (in Htr2c-null mice), we investigate the role of Htr2c in forebrain Sim1 neurons. Unexpectedly, we find that Htr2c acts in these neurons to promote food intake and counteract the anorectic effect of serotonergic appetite suppressants. Furthermore, Htr2c marks a subset of Sim1 neurons in the paraventricular nucleus of the hypothalamus (PVH). Chemogenetic activation of these neurons in adult mice suppresses hunger, whereas their silencing promotes feeding. In support of an orexigenic role of PVH Htr2c, whole-cell patch-clamp experiments demonstrate that activation of Htr2c inhibits PVH neurons. Intriguingly, this inhibition is due to Gαi/o-dependent activation of ATP-sensitive K+ conductance, a mechanism of action not identified previously in the mammalian nervous system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Clozapine N-oxide hydrochloride, ≥98% (HPLC), Water soluble Clozapine N-oxide
Sigma-Aldrich
Glybenclamid, ≥99% (HPLC)
Sigma-Aldrich
Lorcaserin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-Gq/11α-Antikörper, CT, Upstate®, from rabbit