Merck
CRISPR screens unveil signal hubs for nutrient licensing of T cell immunity.
Nature (2021-11-20)
Lingyun Long, Jun Wei, Seon Ah Lim, Jana L Raynor, Hao Shi, Jon P Connelly, Hong Wang, Cliff Guy, Boer Xie, Nicole M Chapman, Guotong Fu, Yanyan Wang, Hongling Huang, Wei Su, Jordy Saravia, Isabel Risch, Yong-Dong Wang, Yuxin Li, Mingming Niu, Yogesh Dhungana, Anil Kc, Peipei Zhou, Peter Vogel, Jiyang Yu, Shondra M Pruett-Miller, Junmin Peng, Hongbo Chi
ZUSAMMENFASSUNG
Nutrients are emerging regulators of adaptive immunity1. Selective nutrients interplay with immunological signals to activate mechanistic target of rapamycin complex 1 (mTORC1), a key driver of cell metabolism2-4, but how these environmental signals are integrated for immune regulation remains unclear. Here we use genome-wide CRISPR screening combined with protein-protein interaction networks to identify regulatory modules that mediate immune receptor- and nutrient-dependent signalling to mTORC1 in mouse regulatory T (Treg) cells. SEC31A is identified to promote mTORC1 activation by interacting with the GATOR2 component SEC13 to protect it from SKP1-dependent proteasomal degradation. Accordingly, loss of SEC31A impairs T cell priming and Treg suppressive function in mice. In addition, the SWI/SNF complex restricts expression of the amino acid sensor CASTOR1, thereby enhancing mTORC1 activation. Moreover, we reveal that the CCDC101-associated SAGA complex is a potent inhibitor of mTORC1, which limits the expression of glucose and amino acid transporters and maintains T cell quiescence in vivo. Specific deletion of Ccdc101 in mouse Treg cells results in uncontrolled inflammation but improved antitumour immunity. Collectively, our results establish epigenetic and post-translational mechanisms that underpin how nutrient transporters, sensors and transducers interplay with immune signals for three-tiered regulation of mTORC1 activity and identify their pivotal roles in licensing T cell immunity and immune tolerance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
KOD Heißstart-DNA-Polymerase, High fidelity DNA polymerase designed for accurate PCR amplification of long strand and GC- rich DNA templates for cloning and cDNA amplification applications.