Merck
Reciprocal keratin 18 Ser48 O-GlcNAcylation and Ser52 phosphorylation using peptide analysis.
Biochemical and biophysical research communications (2006-11-07)
Guo-Zhong Tao, Celeste Kirby, Stephen A Whelan, Frank Rossi, Xiahui Bi, Michael MacLaren, Erik Gentalen, Roger A O'Neill, Gerald W Hart, M Bishr Omary
ZUSAMMENFASSUNG
Phosphorylation and O-GlcNAcylation of keratin 18 (K18) are highly dynamic and involve primarily independent K18 populations. We used in vitro phosphorylation and O-GlcNAcylation of wild-type, phospho-Ser52, glyco-Ser48, and Ser-to-Ala mutant 17mer peptides (K18 amino acids 40-56), which include the major K18 glycosylation (Ser48) and phosphorylation (Ser52) sites, to address whether each modification blocks the other. The glyco-K18 peptide blocks Ser52 phosphorylation by protein kinase C, an in vivo K18 kinase, while the phospho-K18 peptide blocks its O-GlcNAcylation. Our findings support the reciprocity of these two post-translational modifications. Therefore, regulation of protein Ser/Thr phosphorylation and glycosylation at proximal sites can be interdependent and provides a potential mechanism of counter regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PKCε Protein, active, 10 µg, Active, N-terminal His6-tagged, recombinant PKCε, amino acids 2-end. For use in Kinase Assays.