Merck
  • Dissecting the roles of the Tuberin protein in the subcellular localization of the G2/M Cyclin, Cyclin B1.
Dissecting the roles of the Tuberin protein in the subcellular localization of the G2/M Cyclin, Cyclin B1.
PloS one (2022-08-11)
Adam Pillon, Jessica Dare-Shih, Jackie Fong, Elizabeth Fidalgo da Silva, Lisa A Porter
ZUSAMMENFASSUNG
Tuberin is a major component of the protein regulatory complex known as the Tuberous Sclerosis Complex and plays a crucial role in cell cycle progression and protein synthesis. Mutations in the Tuberin gene, TSC2, lead to the formation of benign tumors in many organ systems and causes the Tuberous Sclerosis Complex disorder. Genotypes ranging from point mutations to large deletions in the TSC2 gene have been clinically characterized with a wide range of phenotypes from skin tumors to large brain tumors. Our lab has previously demonstrated that Tuberin can directly bind and regulate the timing of nuclear transport of the G2/M cyclin, Cyclin B1. Herein we study the consequence of one clinically relevant truncation in the Tuberin protein on cell cycle function. We demonstrate that exogenous expression of a fragment of the N-term region of Tuberin alters the subcellular localization of Cyclin B1 and increases cell proliferation. This adds to our body of information about the residues within Tuberin responsible for regulating the cytoplasmic retention of Cyclin B1 and supports the phenotypic data seen in the clinic with Tuberous Sclerosis Complex patients harbouring similar large deletions in Tuberin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution