Merck
  • Nuclear Prospero allows one-division potential to neural precursors and post-mitotic status to neurons via opposite regulation of Cyclin E.
Nuclear Prospero allows one-division potential to neural precursors and post-mitotic status to neurons via opposite regulation of Cyclin E.
PLoS genetics (2022-08-09)
Jordan Mar, Kalpana Makhijani, Denise Flaherty, Krishna Moorthi Bhat
ZUSAMMENFASSUNG
In Drosophila embryonic CNS, the multipotential stem cells called neuroblasts (NBs) divide by self-renewing asymmetric division and generate bipotential precursors called ganglion mother cells (GMCs). GMCs divide only once to generate two distinct post-mitotic neurons. The genes and the pathways that confer a single division potential to precursor cells or how neurons become post-mitotic are unknown. It has been suggested that the homeodomain protein Prospero (Pros) when localized to the nucleus, limits the stem-cell potential of precursors. Here we show that nuclear Prospero is phosphorylated, where it binds to chromatin. In NB lineages such as MP2, or GMC lineages such as GMC4-2a, Pros allows the one-division potential, as well as the post-mitotic status of progeny neurons. These events are mediated by augmenting the expression of Cyclin E in the precursor and repressing the expression in post-mitotic neurons. Thus, in the absence of Pros, Cyclin E is downregulated in the MP2 cell. Consequently, MP2 fails to divide, instead, it differentiates into one of the two progeny neurons. In progeny cells, Pros reverses its role and augments the downregulation of Cyclin E, allowing neurons to exit the cell cycle. Thus, in older pros mutant embryos Cyclin E is upregulated in progeny cells. These results elucidate a long-standing problem of division potential of precursors and post-mitotic status of progeny cells and how fine-tuning cyclin E expression in the opposite direction controls these fundamental cellular events. This work also sheds light on the post-translational modification of Pros that determines its cytoplasmic versus nuclear localization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktailpulver, for general use, lyophilized powder
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Lambda protein phosphatase, liquid, Bacteriophage Lambda, recombinant, expressed in E. coli