Merck
  • Differentiation of human induced pluripotent stem cells into hypothalamic vasopressin neurons with minimal exogenous signals and partial conversion to the naive state.
Differentiation of human induced pluripotent stem cells into hypothalamic vasopressin neurons with minimal exogenous signals and partial conversion to the naive state.
Scientific reports (2022-10-18)
Hajime Ozaki, Hidetaka Suga, Mayu Sakakibara, Mika Soen, Natsuki Miyake, Tsutomu Miwata, Shiori Taga, Takashi Nagai, Mayuko Kano, Kazuki Mitsumoto, Takashi Miyata, Tomoko Kobayashi, Mariko Sugiyama, Takeshi Onoue, Hiroshi Takagi, Daisuke Hagiwara, Shintaro Iwama, Ryoichi Banno, Genzo Iguchi, Yutaka Takahashi, Keiko Muguruma, Haruhisa Inoue, Hiroshi Arima
ZUSAMMENFASSUNG
Familial neurohypophyseal diabetes insipidus (FNDI) is a degenerative disease of vasopressin (AVP) neurons. Studies in mouse in vivo models indicate that accumulation of mutant AVP prehormone is associated with FNDI pathology. However, studying human FNDI pathology in vivo is technically challenging. Therefore, an in vitro human model needs to be developed. When exogenous signals are minimized in the early phase of differentiation in vitro, mouse embryonic stem cells (ESCs)/induced pluripotent stem cells (iPSCs) differentiate into AVP neurons, whereas human ESCs/iPSCs die. Human ESCs/iPSCs are generally more similar to mouse epiblast stem cells (mEpiSCs) compared to mouse ESCs. In this study, we converted human FNDI-specific iPSCs by the naive conversion kit. Although the conversion was partial, we found improved cell survival under minimal exogenous signals and differentiation into rostral hypothalamic organoids. Overall, this method provides a simple and straightforward differentiation direction, which may improve the efficiency of hypothalamic differentiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
DNAse I, from bovine pancreas
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-TRA-1-60-Antikörper, Klon TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic-Antigen-4-Antikörper, Klon MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurophysin 2/NP-AVP Antibody, clone PS 41, clone PS 41, from mouse