Merck
  • Identifying genome-wide off-target sites of CRISPR RNA-guided nucleases and deaminases with Digenome-seq.
Identifying genome-wide off-target sites of CRISPR RNA-guided nucleases and deaminases with Digenome-seq.
Nature protocols (2021-01-20)
Daesik Kim, Beum-Chang Kang, Jin-Soo Kim
ZUSAMMENFASSUNG
Digested genome sequencing (Digenome-seq) is a highly sensitive, easy-to-carry-out, cell-free method for experimentally identifying genome-wide off-target sites of programmable nucleases and deaminases (also known as base editors). Genomic DNA is digested in vitro using clustered regularly interspaced short palindromic repeats ribonucleoproteins (RNPs; plus DNA-modifying enzymes to cleave both strands of DNA at sites containing deaminated base products, in the case of base editors) and subjected to whole-genome sequencing (WGS) with a typical sequencing depth of 30×. A web-based program is available to map in vitro cleavage sites corresponding to on- and off-target sites. Chromatin DNA, in parallel with histone-free genomic DNA, can also be used to account for the effects of chromatin structure on off-target nuclease activity. Digenome-seq is more sensitive and comprehensive than cell-based methods for identifying off-target sites. Unlike other cell-free methods, Digenome-seq does not involve enrichment of DNA ends through PCR amplification. The entire process other than WGS, which takes ~1-2 weeks, including purification and preparation of RNPs, digestion of genomic DNA and bioinformatic analysis after WGS, takes about several weeks.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
RNase A -Lösung