Merck
  • Analysis of Interaction Between Long Noncoding RNAs and Protein by RNA Immunoprecipitation in Arabidopsis.
Analysis of Interaction Between Long Noncoding RNAs and Protein by RNA Immunoprecipitation in Arabidopsis.
Methods in molecular biology (Clifton, N.J.) (2019-04-05)
Jun Sung Seo, Nam-Hai Chua
ZUSAMMENFASSUNG
Long noncoding RNAs (lncRNAs) play important roles in several processes including control of gene expression. These RNAs function through binding to histone-modifying complexes and transcriptional machinery including transcription factor, mediator, and RNA polymerase II. We present methods for the discovery and characterization of lncRNAs. RNA immunoprecipitation (RIP) is a modified version of chromatin immunoprecipitation (ChIP), and it is now generally used in lncRNA study. The method allows for testing of lncRNA-protein interactions in vivo. RIP assay facilitates the identification of consensus sequences of preferred binding site for the RNA-binding protein under study, and identification of the binding sites can provide valuable information on the possible mechanism by which the RNA-binding protein functions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Protein-A-Agarose/Lachssperma-DNA, 2,5 mL, for use in chromatin immunoprecipitations (ChIP assays)