Merck
POGZ promotes homology-directed DNA repair in an HP1-dependent manner.
EMBO reports (2021-11-11)
John Heath, Estelle Simo Cheyou, Steven Findlay, Vincent M Luo, Edgar Pinedo Carpio, Jeesan Lee, Billel Djerir, Xiaoru Chen, Théo Morin, Benjamin Lebeau, Martin Karam, Halil Bagci, Damien Grapton, Josie Ursini-Siegel, Jean-Francois Côté, Michael Witcher, Stéphane Richard, Alexandre Maréchal, Alexandre Orthwein
ZUSAMMENFASSUNG
The heterochromatin protein HP1 plays a central role in the maintenance of genome stability but little is known about how HP1 is controlled. Here, we show that the zinc finger protein POGZ promotes the presence of HP1 at DNA double-strand breaks (DSBs) in human cells. POGZ depletion delays the resolution of DSBs and sensitizes cells to different DNA-damaging agents, including cisplatin and talazoparib. Mechanistically, POGZ promotes homology-directed DNA repair by retaining the BRCA1/BARD1 complex at DSBs in an HP1-dependent manner. In vivo CRISPR inactivation of Pogz is embryonically lethal. Pogz haploinsufficiency (Pogz+ /delta) results in developmental delay, impaired intellectual abilities, hyperactive behaviour and a compromised humoral immune response in mice, recapitulating the main clinical features of the White Sutton syndrome (WHSUS). Pogz+ /delta mice are further radiosensitive and accumulate DSBs in diverse tissues, including the spleen and brain. Altogether, our findings identify POGZ as an important player in homology-directed DNA repair both in vitro and in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Anti-Kaninchen-IgG-Antikörper der Ziege, HRP-konjugiert, 1 mg/mL, Upstate®
Sigma-Aldrich
Anti-Maus-IgG-Antikörper des Kaninchens, HRP-Konjugat, Chemicon®, from rabbit
Sigma-Aldrich
Anti-BRCA1-Antikörper, serum, Upstate®