- Preparation and Application of a Decellularized Extracellular Matrix for Identification of ADAMTS Substrates.
Preparation and Application of a Decellularized Extracellular Matrix for Identification of ADAMTS Substrates.
Methods in molecular biology (Clifton, N.J.) (2019-08-30)
Rahel Schnellmann, Ruth Chiquet-Ehrismann
PMID31463920
ZUSAMMENFASSUNG
Here we describe the use of a decellularized ECM produced in vitro by BALB/c 3T3 fibroblasts for the identification of ADAMTS substrates. Seeding of ADAMTS protease-producing HEK cells on top of the cell-free ECM followed by analysis of the conditioned medium by liquid chromatography tandem mass spectrometry (LC-MS/MS), allows for screening of ADAMTS substrates without prior purification of full-length protease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture