Merck
  • Enhanced maintenance of rat islets of Langerhans on laminin-coated electrospun nanofibrillar matrix in vitro.
Enhanced maintenance of rat islets of Langerhans on laminin-coated electrospun nanofibrillar matrix in vitro.
Cell biology international (2013-02-15)
Mozhdeh Sojoodi, Ali Farrokhi, Azadeh Moradmand, Hossein Baharvand
ZUSAMMENFASSUNG
Understanding the extracellular matrix (ECM) effect on pancreatic β cells is critical to optimise the derivation of functional β cells for transplantation and understand mechanisms that control islet neogenesis and glucose homeostasis. We assessed the effect of natural ECMs [collagen I, collagen IV, laminin and fibronectin (FN)] on rat islets of Langerhans' morphology, adhesion, viability, functionality and islet specific genes expression after 7 days in vitro culture. However, we could not detect a significant difference on the other parameters in these ECMs and islets interaction. To examine islets interactions, we used a synthetic three dimensional surface composed of electrospun polyamide nanofibres. Laminin-coated nanofibrillar surfaces, but not laminin or nanosurface alone, induced comparable expression of the Ins1 and Ins2 genes in adult β cells. Using a glucose challenge test, a marked response of insulin secretion by islets occurred that were cultured on laminin-coated nanofibrillar surfaces.We contend that the reestablishment of cellular interactions by the combination of nanomaterials and natural ECMs can be useful in maintaining in vitro islet functions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibronectin-Humanplasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
4′,6-Diamidin-2-phenylindol -dihydrochlorid, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Diazoxid