Merck
Chromatin immunoprecipitation analysis in filamentous fungi.
Methods in molecular biology (Clifton, N.J.) (2012-10-16)
Stefan Boedi, Yazmid Reyes-Dominguez, Joseph Strauss
ZUSAMMENFASSUNG
Chromatin immunoprecipitation (ChIP) is used to map the interaction between proteins and DNA at a specific genomic locus in the living cell. The protein-DNA complexes are stabilized already in vivo by reversible crosslinking and the DNA is sheared by sonication or enzymatic digestion into fragments suitable for the subsequent immunoprecipitation step. Antibodies recognizing chromatin-linked proteins, transcription factors, artificial tags, or specific protein modifications are then used to pull down DNA-protein complexes containing the target. After reversal of crosslinks and DNA purification locus-specific quantitative PCR is used to determine the amount of DNA that was associated with the target at a given time point and experimental condition. DNA quantification can be carried out for several genomic regions by multiple qPCRs or at a genome-wide scale by massive parallel sequencing (ChIP-Seq).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Acetyl-Histon-H3-Antikörper, from rabbit
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys14)-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Acetyl-Histon-H4-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Dimethylhiston-H3-(Lys4)-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-Histon H3-Antikörper, 0.5 mg/mL, Upstate®