Merck
The linker histone H1.2 is a novel component of the nucleolar organizer regions.
The Journal of biological chemistry (2018-01-06)
Junjie Chen, Boon Heng Dennis Teo, Yitian Cai, Seng Yin Kelly Wee, Jinhua Lu
ZUSAMMENFASSUNG
The nucleoli accumulate rRNA genes and are the sites of rRNA synthesis and rRNA assembly into ribosomes. During mitosis, nucleoli dissociate, but nucleolar remnants remain on the rRNA gene loci, forming distinct nucleolar organizer regions (NORs). Little is known about the composition and structure of NORs, but upstream binding factor (UBF) has been established as its master organizer. In this study, we sought to establish new proteins in NORs. Using UBF-Sepharose to isolate UBF-binding proteins, we identified histone H1.2 as a candidate partner but were puzzled by this observation, given that UBF is known to be located predominantly in nucleoli, whereas H1.2 distributed broadly among the chromatins in interphase nuclei. We then examined cells undergoing mitosis and saw that both H1.2 and UBF were recruited into NORs in this state, reconciling the results of our UBF pulldowns. Inhibiting rRNA synthesis in interphase nuclei also induced NOR-like structures containing both UBF and H1.2. When chromosomes were isolated and spread on coverslips, NORs appeared separated from the chromosomes containing both UBF and H1.2. After chromosomes were fragmented by homogenization, intact NORs remained visible. Results collectively suggest that NORs are independent structures and that the linker histone H1.2 is a novel component of this structure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-RNA-Polymerase II-Antikörper, Klon CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Anti-B23 antibody, Mouse monoclonal, clone FC82291, purified from hybridoma cell culture