Merck
Knocking out alpha-synuclein in melanoma cells downregulates L1CAM and decreases motility.
Scientific reports (2023-06-08)
Nithya Gajendran, Santhanasabapathy Rajasekaran, Stephan N Witt
ZUSAMMENFASSUNG
The Parkinson's disease (PD) associated protein, alpha-synuclein (α-syn/SNCA), is highly expressed in aggressive melanomas. The goal of this study was to reveal possible mechanism(s) of α-syn involvement in melanoma pathogenesis. Herein, we asked whether α-syn modulates the expression of the pro-oncogenic adhesion molecules L1CAM and N-cadherin. We used two human melanoma cell lines (SK-MEL-28, SK-MEL-29), SNCA-knockout (KO) clones, and two human SH-SY5Y neuroblastoma cell lines. In the melanoma lines, loss of α-syn expression resulted in significant decreases in the expression of L1CAM and N-cadherin and concomitant significant decreases in motility. On average, there was a 75% reduction in motility in the four SNCA-KOs tested compared to control cells. Strikingly, comparing neuroblastoma SH-SY5Y cells that have no detectable α-syn to SH-SY5Y cells that stably express α-syn (SH/+αS), we found that expressing α-syn increased L1CAM and single-cell motility by 54% and 597%, respectively. The reduction in L1CAM level in SNCA-KO clones was not due to a transcriptional effect, rather we found that L1CAM is more efficiently degraded in the lysosome in SNCA-KO clones than in control cells. We propose that α-syn is pro-survival to melanoma (and possibly neuroblastoma) because it promotes the intracellular trafficking of L1CAM to the plasma membrane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bafilomycin A1 aus Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid