Merck
NKT cells adopt a glutamine-addicted phenotype to regulate their homeostasis and function.
Cell reports (2022-10-27)
Ajay Kumar, Emily L Yarosz, Anthony Andren, Li Zhang, Costas A Lyssiotis, Cheong-Hee Chang
ZUSAMMENFASSUNG
Natural killer T (NKT) cells operate distinctly different metabolic programming from CD4 T cells, including a strict requirement for glutamine to regulate cell homeostasis. However, the underlying mechanisms remain unknown. Here, we report that at a steady state, NKT cells have higher glutamine levels than CD4 T cells and that NKT cells increase glutaminolysis on activation. Activated NKT cells use glutamine to fuel the tricarboxylic acid cycle and glutathione synthesis. In addition, glutamine-derived nitrogen enables protein glycosylation via the hexosamine biosynthesis pathway (HBP). Each of these branches of glutamine metabolism seems to be critical for NKT cell homeostasis and mitochondrial functions. Glutaminolysis and HBP differentially regulate interleukin-4 (IL-4) and interferon γ (IFNγ) production. Glutamine metabolism appears to be controlled by AMP-activated protein kinase (AMPK)-mammalian target of rapamycin complex 1 (mTORC1) signaling. These findings highlight a distinct metabolic requirement of NKT cells compared with CD4 T cells, which may have therapeutic implications in the treatment of certain nutrient-restricted diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Antimycin A aus Streptomyces sp.
Sigma-Aldrich
Carbonylcyanid-4-(trifluormethoxy)phenylhydrazon, ≥98% (HPLC), powder
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Rotenon, ≥95%
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
(−)-Epigallokatechingallat, ≥95%
Sigma-Aldrich
L-Buthioninsulfoximin, ≥97% (TLC)