Merck
  • Cancer-associated fibroblasts promote tumor cell growth via miR-493-5p in intrahepatic cholangiocarcinoma.
Cancer-associated fibroblasts promote tumor cell growth via miR-493-5p in intrahepatic cholangiocarcinoma.
Cancer science (2022-11-13)
Katsuya Toshida, Shinji Itoh, Noboru Harada, Akinari Morinaga, Kyohei Yugawa, Takahiro Tomiyama, Yukiko Kosai-Fujimoto, Takahiro Tomino, Takeshi Kurihara, Yoshihiro Nagao, Kazutoyo Morita, Yoshinao Oda, Tomoharu Yoshizumi
ZUSAMMENFASSUNG
The association between tumor microenvironment (TME) and cancer-associated fibroblasts (CAFs) in intrahepatic cholangiocarcinoma (ICC) progression is poorly understood. This study aimed to reveal whether specific microRNAs (miRNAs) in extracellular vesicles (EVs) derived from CAFs were involved in ICC progression. Conditioned medium (CM) and EVs in the CM of CAFs and normal fibroblasts (NFs) derived from ICC specimens were used to investigate the effects on tumor cell lines. miRNA microarray assay was used to examine the miRNAs of EVs derived from CAFs and NFs in ICC, and the effects of miR-493-5p on tumor cell lines were examined. Additionally, databases were used to identify miR-493-5p targets, and the relationship between prognosis of ICC patients and cocaine- and amphetamine-regulated transcript propeptide (CARTPT), one of the targets of miR-493-5p, expression in ICC tissues was retrospectively analyzed. Compared with NF-derived CM and EVs, CAF-derived CM and EVs promoted cell lines in proliferation, scratch, migration, and invasion assays. miRNA microarray analysis revealed that miR-493-5p was significantly increased in CAF-derived EVs compared to NF-derived EVs. Tumor cell lines transfected with miR-493-5p were promoted in proliferation and scratch assays. Immunohistochemical staining was performed on 76 ICC specimens; both overall and recurrence-free survival rates were significantly worse in the CARTPT-negative group. Univariate and multivariate analyses showed that low CARTPT expression was an independent poor prognostic factor for overall and recurrence-free survival. Overall, our data suggest that CAFs in the ICC TME suppress CARTPT in tumor cells and promote tumor cells via miR-493-5p in EVs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti--α-Glattmuskel-Aktin-Antikörper, Maus, monoklonal, clone 1A4, purified from hybridoma cell culture