Merck
  • TRIM71 reactivation enhances the mitotic and hair cell-forming potential of cochlear supporting cells.
TRIM71 reactivation enhances the mitotic and hair cell-forming potential of cochlear supporting cells.
EMBO reports (2023-07-26)
Xiao-Jun Li, Charles Morgan, Prathamesh T Nadar-Ponniah, Waldemar Kolanus, Angelika Doetzlhofer
ZUSAMMENFASSUNG
Cochlear hair cell loss is a leading cause of deafness in humans. Neighboring supporting cells have some capacity to regenerate hair cells. However, their regenerative potential sharply declines as supporting cells undergo maturation (postnatal day 5 in mice). We recently reported that reactivation of the RNA-binding protein LIN28B restores the hair cell-regenerative potential of P5 cochlear supporting cells. Here, we identify the LIN28B target Trim71 as a novel and equally potent enhancer of supporting cell plasticity. TRIM71 is a critical regulator of stem cell behavior and cell reprogramming; however, its role in cell regeneration is poorly understood. Employing an organoid-based assay, we show that TRIM71 re-expression increases the mitotic and hair cell-forming potential of P5 cochlear supporting cells by facilitating their de-differentiation into progenitor-like cells. Our mechanistic work indicates that TRIM71's RNA-binding activity is essential for such ability, and our transcriptomic analysis identifies gene modules that are linked to TRIM71 and LIN28B-mediated supporting cell reprogramming. Furthermore, our study uncovers that the TRIM71-LIN28B target Hmga2 is essential for supporting cell self-renewal and hair cell formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Phosphatasehemmer-Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatasehemmer-Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Valproinsäure Natriumsalz, 98%
Sigma-Aldrich
LY-411575, ≥97% (HPLC)