Merck
  • Activation of HERV-K(HML-2) disrupts cortical patterning and neuronal differentiation by increasing NTRK3.
Activation of HERV-K(HML-2) disrupts cortical patterning and neuronal differentiation by increasing NTRK3.
Cell stem cell (2021-05-06)
Vidya Padmanabhan Nair, Hengyuan Liu, Gabriele Ciceri, Johannes Jungverdorben, Goar Frishman, Jason Tchieu, Gustav Y Cederquist, Ina Rothenaigner, Kenji Schorpp, Lena Klepper, Ryan M Walsh, Tae Wan Kim, Daniela Cornacchia, Andreas Ruepp, Jens Mayer, Kamyar Hadian, Dmitrij Frishman, Lorenz Studer, Michelle Vincendeau
ZUSAMMENFASSUNG
The biological function and disease association of human endogenous retroviruses (HERVs) are largely elusive. HERV-K(HML-2) has been associated with neurotoxicity, but there is no clear understanding of its role or mechanistic basis. We addressed the physiological functions of HERV-K(HML-2) in neuronal differentiation using CRISPR engineering to activate or repress its expression levels in a human-pluripotent-stem-cell-based system. We found that elevated HERV-K(HML-2) transcription is detrimental for the development and function of cortical neurons. These effects are cell-type-specific, as dopaminergic neurons are unaffected. Moreover, high HERV-K(HML-2) transcription alters cortical layer formation in forebrain organoids. HERV-K(HML-2) transcriptional activation leads to hyperactivation of NTRK3 expression and other neurodegeneration-related genes. Direct activation of NTRK3 phenotypically resembles HERV-K(HML-2) induction, and reducing NTRK3 levels in context of HERV-K(HML-2) induction restores cortical neuron differentiation. Hence, these findings unravel a cell-type-specific role for HERV-K(HML-2) in cortical neuron development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N6,2′-O-Dibutyryladenosin 3′,5′-zyklisches Monophosphat Natriumsalz Monohydrat, ≥96% (HPLC), powder
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-SATB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Tbr2-Antikörper, from rabbit, purified by affinity chromatography