Merck
  • Heterogeneity of perivascular astrocyte endfeet depending on vascular regions in the mouse brain.
Heterogeneity of perivascular astrocyte endfeet depending on vascular regions in the mouse brain.
iScience (2023-10-13)
Takeshi Kameyama, Muneaki Miyata, Hajime Shiotani, Jun Adachi, Soichiro Kakuta, Yasuo Uchiyama, Kiyohito Mizutani, Yoshimi Takai
ZUSAMMENFASSUNG
Astrocytes interact with not only synapses but also brain blood vessels through perivascular astrocyte endfeet (PV-AEF) to form the neurovascular unit (NVU). However, PV-AEF components have not been fully identified. Here, we biochemically isolated blood vessels from mouse brain homogenates and purified PV-AEF. The purified PV-AEF were observed in different sizes, similar to PV-AEF on brain blood vessels. Mass spectrometry analysis identified 9,762 proteins in the purified PV-AEF, including cell adhesion molecules, nectin-2δ, Kirrel2, and podoplanin. Immunofluorescence microscopic analysis revealed that nectin-2δ and podoplanin were concentrated mainly in arteries/arterioles and veins/venules of the mouse brain, whereas Kirrel2 was mainly in arteries/arterioles. Nectin-2α/δ, Kirrel2, and podoplanin were preferentially observed in large sizes of the purified PV-AEF. Furthermore, Kirrel2 potentially has cell adhesion activity of cultured astrocytes. Collectively, these results indicate that PV-AEF have heterogeneity in sizes and molecular components, implying different roles of PV-AEF in NVU function depending on vascular regions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti--α-Glattmuskel-Aktin-Antikörper, Maus, monoklonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal, clone 4H8-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Guinea Pig IgG (H+L), highly cross-adsorbed, CF555 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution