Merck
  • Gestational diabetes in mice induces hematopoietic memory that affects the long-term health of the offspring.
Gestational diabetes in mice induces hematopoietic memory that affects the long-term health of the offspring.
The Journal of clinical investigation (2023-11-21)
Vinothini Govindarajah, Masahide Sakabe, Samantha Good, Michael Solomon, Ashok Arasu, Nong Chen, Xuan Zhang, H Leighton Grimes, Ady Kendler, Mei Xin, Damien Reynaud
ZUSAMMENFASSUNG
Gestational diabetes is a common medical complication of pregnancy that is associated with adverse perinatal outcomes and an increased risk of metabolic diseases and atherosclerosis in adult offspring. The mechanisms responsible for this delayed pathological transmission remain unknown. In mouse models, we found that the development of atherosclerosis in adult offspring born to diabetic pregnancy can be in part linked to hematopoietic alterations. Although they do not show any gross metabolic disruptions, the adult offspring maintain hematopoietic features associated with diabetes, indicating the acquisition of a lasting diabetic hematopoietic memory. We show that the induction of this hematopoietic memory during gestation relies on the activity of the advanced glycation end product receptor (AGER) and the nucleotide binding and oligomerization domain-like receptor family pyrin domain-containing 3 (NLRP3) inflammasome, which lead to increased placental inflammation. In adult offspring, we find that this memory is associated with DNA methyltransferase 1 (DNMT1) upregulation and epigenetic changes in hematopoietic progenitors. Together, our results demonstrate that the hematopoietic system can acquire a lasting memory of gestational diabetes and that this memory constitutes a pathway connecting gestational health to adult pathologies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-FOXO3A Antibody, from rabbit, purified by affinity chromatography