Merck
  • NRF2 activation induces NADH-reductive stress, providing a metabolic vulnerability in lung cancer.
NRF2 activation induces NADH-reductive stress, providing a metabolic vulnerability in lung cancer.
Cell metabolism (2023-02-26)
Tommy Weiss-Sadan, Maolin Ge, Makiko Hayashi, Magdy Gohar, Cong-Hui Yao, Adriaan de Groot, Stefan Harry, Alexander Carlin, Hannah Fischer, Lei Shi, Ting-Yu Wei, Charles H Adelmann, Konstantin Wolf, Tristan Vornbäumen, Benedikt R Dürr, Mariko Takahashi, Marianne Richter, Junbing Zhang, Tzu-Yi Yang, Vindhya Vijay, David E Fisher, Aaron N Hata, Marcia C Haigis, Raul Mostoslavsky, Nabeel Bardeesy, Thales Papagiannakopoulos, Liron Bar-Peled
ZUSAMMENFASSUNG
Multiple cancers regulate oxidative stress by activating the transcription factor NRF2 through mutation of its negative regulator, KEAP1. NRF2 has been studied extensively in KEAP1-mutant cancers; however, the role of this pathway in cancers with wild-type KEAP1 remains poorly understood. To answer this question, we induced NRF2 via pharmacological inactivation of KEAP1 in a panel of 50+ non-small cell lung cancer cell lines. Unexpectedly, marked decreases in viability were observed in >13% of the cell lines-an effect that was rescued by NRF2 ablation. Genome-wide and targeted CRISPR screens revealed that NRF2 induces NADH-reductive stress, through the upregulation of the NAD+-consuming enzyme ALDH3A1. Leveraging these findings, we show that cells treated with KEAP1 inhibitors or those with endogenous KEAP1 mutations are selectively vulnerable to Complex I inhibition, which impairs NADH oxidation capacity and potentiates reductive stress. Thus, we identify reductive stress as a metabolic vulnerability in NRF2-activated lung cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Rotenon, ≥95%
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution