Merck
  • Large-scale phosphomimetic screening identifies phospho-modulated motif-based protein interactions.
Large-scale phosphomimetic screening identifies phospho-modulated motif-based protein interactions.
Molecular systems biology (2023-05-23)
Johanna Kliche, Dimitriya Hristoforova Garvanska, Leandro Simonetti, Dilip Badgujar, Doreen Dobritzsch, Jakob Nilsson, Norman E Davey, Ylva Ivarsson
ZUSAMMENFASSUNG
Phosphorylation is a ubiquitous post-translation modification that regulates protein function by promoting, inhibiting or modulating protein-protein interactions. Hundreds of thousands of phosphosites have been identified but the vast majority have not been functionally characterised and it remains a challenge to decipher phosphorylation events modulating interactions. We generated a phosphomimetic proteomic peptide-phage display library to screen for phosphosites that modulate short linear motif-based interactions. The peptidome covers ~13,500 phospho-serine/threonine sites found in the intrinsically disordered regions of the human proteome. Each phosphosite is represented as wild-type and phosphomimetic variant. We screened 71 protein domains to identify 248 phosphosites that modulate motif-mediated interactions. Affinity measurements confirmed the phospho-modulation of 14 out of 18 tested interactions. We performed a detailed follow-up on a phospho-dependent interaction between clathrin and the mitotic spindle protein hepatoma-upregulated protein (HURP), demonstrating the essentiality of the phospho-dependency to the mitotic function of HURP. Structural characterisation of the clathrin-HURP complex elucidated the molecular basis for the phospho-dependency. Our work showcases the power of phosphomimetic ProP-PD to discover novel phospho-modulated interactions required for cellular function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nocodazol, ≥99% (TLC), powder
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)