Merck
Unique transducins expressed in long and short photoreceptors of lamprey Petromyzon marinus.
Vision research (2008-08-09)
Hakim Muradov, Vasily Kerov, Kimberly K Boyd, Nikolai O Artemyev
ZUSAMMENFASSUNG
Lampreys represent the most primitive vertebrate class of jawless fish and serve as an evolutionary model of the vertebrate visual system. Transducin-alpha (G alpha(t)) subunits were investigated in lamprey Petromyzon marinus in order to understand the molecular origins of rod and cone photoreceptor G proteins. Two G alpha(t) subunits, G alpha(tL) and G alpha(tS), were identified in the P. marinus retina. G alpha(tL) is equally distant from cone and rod G proteins and is expressed in the lamprey's long photoreceptors. The short photoreceptor G alpha(tS) is a rod-like transducin-alpha that retains several unique features of cone transducins. Thus, the duplication of the ancestral transducin gene giving rise to rod transducins has already occurred in the last common ancestor of the jawed and jawless vertebrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Hämocyanin aus Megathura crenulata (Napfschnecke)
Sigma-Aldrich
Peroxidase aus Meerrettich, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Peroxidase aus Meerrettich, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
Hämocyanin aus Megathura crenulata (Napfschnecke), PBS solution
Sigma-Aldrich
Peroxidase aus Meerrettich, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type X, ammonium sulfate suspension
Sigma-Aldrich
Peroxidase aus Meerrettich, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Hemocyanin from mollusk, buffered aqueous solution