Merck
Neural extracellular matrix regulates visual sensory motor integration.
iScience (2024-02-06)
Jacqueline Reinhard, Cornelius Mueller-Buehl, Susanne Wiemann, Lars Roll, Veronika Luft, Hamed Shabani, Daniel L Rathbun, Lin Gan, Chao-Chung Kuo, Julia Franzen, Stephanie C Joachim, Andreas Faissner
ZUSAMMENFASSUNG
Visual processing depends on sensitive and balanced synaptic neurotransmission. Extracellular matrix proteins in the environment of cells are key modulators in synaptogenesis and synaptic plasticity. In the present study, we provide evidence that the combined loss of the four extracellular matrix components, brevican, neurocan, tenascin-C, and tenascin-R, in quadruple knockout mice leads to severe retinal dysfunction and diminished visual motion processing in vivo. Remarkably, impaired visual motion processing was accompanied by a developmental loss of cholinergic direction-selective starburst amacrine cells. Additionally, we noted imbalance of inhibitory and excitatory synaptic signaling in the quadruple knockout retina. Collectively, the study offers insights into the functional importance of four key extracellular matrix proteins for retinal function, visual motion processing, and synaptic signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Anti-Digoxigenin-AP, Fab-Fragmente, from sheep
Sigma-Aldrich
Proteasehemmer-Cocktailpulver, for general use, lyophilized powder
Sigma-Aldrich
Anti-Cholin-Acetyltransferase-Antikörper, serum, Chemicon®
Sigma-Aldrich
Anti-VGLUT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution