Merck
The BICD2 dynein cargo adaptor binds to the HPV16 L2 capsid protein and promotes HPV infection.
PLoS pathogens (2024-06-03)
Kaitlyn Speckhart, Jeongjoon Choi, Daniel DiMaio, Billy Tsai
ZUSAMMENFASSUNG
During entry, human papillomavirus (HPV) traffics from the endosome to the trans Golgi network (TGN) and Golgi and then the nucleus to cause infection. Although dynein is thought to play a role in HPV infection, how this host motor recruits the virus to support infection and which entry step(s) requires dynein are unclear. Here we show that the dynein cargo adaptor BICD2 binds to the HPV L2 capsid protein during entry, recruiting HPV to dynein for transport of the virus along the endosome-TGN/Golgi axis to promote infection. In the absence of BICD2 function, HPV accumulates in the endosome and TGN and infection is inhibited. Cell-based and in vitro binding studies identified a short segment near the C-terminus of L2 that can directly interact with BICD2. Our results reveal the molecular basis by which the dynein motor captures HPV to promote infection and identify this virus as a novel cargo of the BICD2 dynein adaptor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
γγ-Sekretase-Inhibitor XXI, Verbindung E, This g-secretase inhibitor, CAS 209986-17-4, is a cell-permeable, potent, selective, non-transition-state analog inhibitor of γ-secretase and Notch processing. Lowers Aβ levels in APP transgenic mice
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-BICDL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-BICD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution