Merck
Dual IKZF2 and CK1α degrader targets acute myeloid leukemia cells.
Cancer cell (2023-03-11)
Sun-Mi Park, David K Miyamoto, Grace Y Q Han, Mandy Chan, Nicole M Curnutt, Nathan L Tran, Anthony Velleca, Jun Hyun Kim, Alexandra Schurer, Kathryn Chang, Wenqing Xu, Michael G Kharas, Christina M Woo
ZUSAMMENFASSUNG
Acute myeloid leukemia (AML) is a hematologic malignancy for which several epigenetic regulators have been identified as therapeutic targets. Here we report the development of cereblon-dependent degraders of IKZF2 and casein kinase 1α (CK1α), termed DEG-35 and DEG-77. We utilized a structure-guided approach to develop DEG-35 as a nanomolar degrader of IKZF2, a hematopoietic-specific transcription factor that contributes to myeloid leukemogenesis. DEG-35 possesses additional substrate specificity for the therapeutically relevant target CK1α, which was identified through unbiased proteomics and a PRISM screen assay. Degradation of IKZF2 and CK1α blocks cell growth and induces myeloid differentiation in AML cells through CK1α-p53- and IKZF2-dependent pathways. Target degradation by DEG-35 or a more soluble analog, DEG-77, delays leukemia progression in murine and human AML mouse models. Overall, we provide a strategy for multitargeted degradation of IKZF2 and CK1α to enhance efficacy against AML that may be expanded to additional targets and indications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lenalidomide, ≥98% (HPLC)
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)
Sigma-Aldrich
Anti-CRBN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution