Merck
Knockout of the regulatory factor X1 gene leads to early embryonic lethality.
Biochemical and biophysical research communications (2009-06-30)
Chenzhuo Feng, Wenhao Xu, Zhiyi Zuo
ZUSAMMENFASSUNG
The biological function of regulatory factor X1 (RFX1), the prototype member of the transcription factor RFX family, is not clear. We have used gene trap technique to disrupt the expression of RFX1 in mice. Although, heterozygous RFX1(+/-) mice appear normal and fertile, homozygous RFX1(-/-) embryos died at an early stage (most likely before embryonic day 2.5). Our results indicate that RFX1 regulates expression of genes that are essential for early embryonic development/survival and that RFX1 function can not be compensated by other RFX1 family members.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Schaftestes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase aus Streptomyces hyalurolyticus
Sigma-Aldrich
Hyaluronidase aus Schaftestes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid