Merck
  • Insulin receptor signaling regulates actin cytoskeletal organization in developing photoreceptors.
Insulin receptor signaling regulates actin cytoskeletal organization in developing photoreceptors.
Journal of neurochemistry (2009-07-07)
Raju V S Rajala, Ammaji Rajala, Richard S Brush, Nora P Rotstein, Luis E Politi
ZUSAMMENFASSUNG
The insulin receptor (IR) and IR signaling proteins are widely distributed throughout the CNS. IR signaling provides a trophic signal for transformed retinal neurons in culture and we recently reported that deletion of IR in rod photoreceptors by Cre/lox system resulted in stress-induced photoreceptor degeneration. These studies suggest a neuroprotective role of IR in rod photoreceptor cell function. However, there are no studies available on the role of insulin-induced IR signaling in the development of normal photoreceptors. To examine the role of insulin-induced IR signaling, we analyzed cultured neuronal cells isolated from newborn rodent retinas. In insulin-lacking cultures, photoreceptors from wild-type rat retinas exhibited an abnormal morphology with a wide axon cone and disorganization of the actin and tubulin cytoskeleton. Photoreceptors from IR knockout mouse retinas also exhibited a similar abnormal morphology. A novel finding in this study was that addition of docosahexaenoic acid, a photoreceptor trophic factor, restored normal axonal outgrowth in insulin-lacking cultures. These data suggest that IR signaling pathways regulate actin and tubulin cytoskeletal organization in photoreceptors; they also imply that insulin and docosahexaenoic acid activate at least partially overlapping signaling pathways that are essential for the development of normal photoreceptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hanks-Salze -Lösung, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks-Salze -Lösung, Modified, with sodium bicarbonate, without phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks-Salze -Lösung, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks-Salze -Lösung, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
Hanks-Salze -Lösung, HBSS Modified, with calcium, with magnesium, without phenol red, liquid, suitable for cell culture
SAFC
Hanks-Salze -Lösung, HBSS Modified, with phenol red, without calcium, without magnesium, liquid
Sigma-Aldrich
Hanks-Salze -Lösung, 10 ×, Modified, without calcium, magnesium or sodium bicarbonate