Merck
  • MPL mutations in essential thrombocythemia uncover a common path of activation with eltrombopag dependent on W491.
MPL mutations in essential thrombocythemia uncover a common path of activation with eltrombopag dependent on W491.
Blood (2020-01-25)
Gabriel Levy, Serge Carillo, Benjamin Papoular, Bruno Cassinat, Jean-Marc Zini, Emilie Leroy, Leila N Varghese, Ilyas Chachoua, Jean-Philippe Defour, Steven O Smith, Stefan N Constantinescu
ZUSAMMENFASSUNG
Mutations in the MPL gene encoding the human thrombopoietin receptor (TpoR) drive sporadic and familial essential thrombocythemias (ETs). We identified 2 ET patients harboring double mutations in cis in MPL, namely, L498W-H499C and H499Y-S505N. Using biochemical and signaling assays along with partial saturation mutagenesis, we showed that L498W is an activating mutation potentiated by H499C and that H499C and H499Y enhance the activity of the canonical S505N mutation. L498W and H499C can activate a truncated TpoR mutant, which lacks the extracellular domain, indicating these mutations act on the transmembrane (TM) cytosolic domain. Using a protein complementation assay, we showed that L498W and H499C strongly drive dimerization of TpoR. Activation by tryptophan substitution is exquisitely specific for position 498. Using structure-guided mutagenesis, we identified upstream amino acid W491 as a key residue required for activation by L498W or canonical activating mutations such as S505N and W515K, as well as by eltrombopag. Structural data point to a common dimerization and activation path for TpoR via its TM domain that is shared between the small-molecule agonist eltrombopag and canonical and novel activating TpoR mutations that all depend on W491, a potentially accessible extracellular residue that could become a target for therapeutic intervention.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Eltrombopag olamine, ≥98% (HPLC)