Merck
Fed-batch two-phase production of alanine by a metabolically engineered Escherichia coli.
Biotechnology letters (2006-08-12)
Geoffrey M Smith, Sarah A Lee, Kevin C Reilly, Mark A Eiteman, Elliot Altman
ZUSAMMENFASSUNG
DL-Alanine was produced from glucose in an Escherichia coli pfl pps poxB ldhA aceEF pTrc99A-alaD strain which lacked pyruvate-formate lyase, phosphoenolpyruvate (PEP) synthase, pyruvate oxidase, lactate dehydogenase, components of the pyruvate dehydogenase complex and over-produced alanine dehydrogenase (ALD). A two-phase process was developed with cell growth under aerobic conditions followed by alanine production under anaerobic conditions. Using the batch mode, cells grew to 5.3 g/l in 9 h with the accumulation of 6-10 g acetate/l, and under subsequent anaerobic conditions achieved 34 g alanine/l in 13 h with a yield of 0.86 g/g glucose. Using the fed-batch mode at micro = 0.15 h(-1), only about 1 g acetate/l formed in the 25 h required for the cells to reach 5.6 g/l, and 88 g alanine/l accumulated during the subsequent 23 h. This fed-batch process attained an alanine volumetric productivity of 4 g/lh during the production phase, and a yield that was essentially 1 g/g.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Alanin-Dehydrogenase aus Bacillus subtilis, buffered aqueous glycerol solution, ~30 units/mg protein (Lowry)
Sigma-Aldrich
Alanin-Dehydrogenase, rekombinant, recombinant, expressed in E. coli, ≥15 U/mg
Sigma-Aldrich
L-Alanin-Dehydrogenase aus Bacillus subtilis, ammonium sulfate suspension, ≥20 units/mg protein (Lowry)