Merck
  • A novel malate dehydrogenase from Ceratonia siliqua L. seeds with potential biotechnological applications.
A novel malate dehydrogenase from Ceratonia siliqua L. seeds with potential biotechnological applications.
The protein journal (2012-09-12)
Clelia Muccio, Vincenzo Guida, Amalia Di Petrillo, Valeria Severino, Antimo Di Maro
ZUSAMMENFASSUNG
A novel malate dehydrogenase (MDH; EC 3.1.1.1.37), hereafter MDHCs, from Ceratonia siliqua seeds, commonly known as Carob tree, was purified by using ammonium sulphate precipitation, ion exchange chromatography on SteamLine SP and gel-filtration. The molecular mass of the native protein, obtained by analytical gel-filtration, was about 65 kDa, whereas, by using SDS-PAGE analysis, with and without reducing agent, was 34 kDa. The specific activity of purified MDHCs (0.25 mg/100 g seeds) was estimated to be 188 U/mg. The optimum activity of the enzyme is at pH 8.5, showing a decrease in the presence of Ca(2+), Mg(2+) and NaCl. The N-terminal sequence of the first 20 amino acids of MDHCs revealed 95 % identity with malate dehydrogenase from Medicago sativa L. Finally, the enzymatic activity of MDHCs was preserved even after absorption onto a PVDF membrane. To our knowledge, this is the first contribution to the characterization of an enzyme from Carob tree sources.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Schweineherz, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Schweineherz, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Rinderherz, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Schweineherz, ≥400 units/mg protein (biuret), ammonium sulfate suspension