Merck
  • Bioactive hyaluronan fragment (hexasaccharide) detects specific hexa-binding proteins in human breast and stomach cancer: possible role in tumorogenesis.
Bioactive hyaluronan fragment (hexasaccharide) detects specific hexa-binding proteins in human breast and stomach cancer: possible role in tumorogenesis.
Indian journal of biochemistry & biophysics (2012-10-20)
Prashanth Srinivas, Srinivas Prasad Kollapalli, Anil Thomas, Karuna Kumar Mortha, Shib Das Banerjee
ZUSAMMENFASSUNG
Hyaluronan (HA) is a component of extracellular matrix that influences cell-proliferation, migration, development, regeneration, normal tissue remodeling, tissues undergoing malignancy and tumor cell interaction. The widespread occurrence of HA binding proteins, their involvement in tissue organization and the control of cellular behavior are well documented. The low molecular mass HA fragments can also induce a variety of biological events, including chemokine gene expression, transcription factor expression and angiogenesis. It is believed that these fragments are more potent in cellular activities than high molecular mass HA. In this study, we isolated the various fragments by gel permeation chromatography of hyaluronidase digested HA and characterized by fluoro assisted carbohydrate electrophoresis (FACE) and matrix assisted laser desorption ionization analysis (MALDI). Detection and distribution of cellular receptors in invasive tumor tissues for HA polymer and HA fragments were determined both by Western blot and histochemistry. The study demonstrated the overexpression of HA-hexa binding protein in human tumors of breast and stomach and its involvement in tumorogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Schaftestes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase aus Schaftestes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid