- FISH analysis using PPAR γ-specific probes for detection of PAX8-PPAR γ translocation in follicular thyroid neoplasms.
FISH analysis using PPAR γ-specific probes for detection of PAX8-PPAR γ translocation in follicular thyroid neoplasms.
Methods in molecular biology (Clifton, N.J.) (2012-10-27)
Noor Akmal Syed Husain Sharifah, Zubaidah Zakaria, Wai Kit Chia
PMID23100233
ZUSAMMENFASSUNG
Fluorescence in situ hybridization (FISH) is increasingly gaining importance in clinical diagnostics settings. Due to the ability of the technique to detect chromosomal abnormalities in samples with low cellularity or containing a mixed population of cells even at a single-cell level, it has become more popular in cancer research and diagnosis. Here, we describe the FISH technique for detection of PAX8-PPARγ translocation in follicular thyroid neoplasms, and the optimal protocol for the detection of this fusion gene using in archival formalin-fixed paraffin-embedded (FFPE) thyroid tissue sections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid