Merck
  • Conformational analysis of StrH, the surface-attached exo-β-D-N-acetylglucosaminidase from Streptococcus pneumoniae.
Conformational analysis of StrH, the surface-attached exo-β-D-N-acetylglucosaminidase from Streptococcus pneumoniae.
Journal of molecular biology (2012-11-17)
Benjamin Pluvinage, Seth Chitayat, Elizabeth Ficko-Blean, D Wade Abbott, Jobby Maroor Kunjachen, Julie Grondin, Holly L Spencer, Steven P Smith, Alisdair B Boraston
ZUSAMMENFASSUNG
Streptococcus pneumoniae is a serious human pathogen that presents on its surface numerous proteins involved in the host-bacterium interaction. The carbohydrate-active enzymes are particularly well represented among these surface proteins, and many of these are known virulence factors, highlighting the importance of carbohydrate processing by this pathogen. StrH is a surface-attached exo-β-D-N-acetylglucosaminidase that cooperates with the sialidase NanA and the β-galactosidase BgaA to sequentially degrade the nonreducing terminal arms of complex N-linked glycans. This enzyme is a large multi-modular protein that is notable for its tandem N-terminal family GH20 catalytic modules, whose individual X-ray crystal structures were recently reported. StrH also contains C-terminal tandem G5 modules, which are uncharacterized. Here, we report the NMR-determined solution structure of the first G5 module in the tandem, G5-1, which along with the X-ray crystal structures of the GH20 modules was used in conjunction with small-angle X-ray scattering to construct a pseudo-atomic model of full-length StrH. The results reveal a model in which StrH adopts an elongated conformation that may project the catalytic modules away from the surface of the bacterium to a distance of up to ~250 Å.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-N-Acetylglucosaminidase aus Canavalia ensiformis (Jack bean), ammonium sulfate suspension, ≥8 units/mg protein
Sigma-Aldrich
β-N-Acetylglucosaminidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution