Merck
  • Responses of B-esterase enzymes in oysters (Crassostrea gigas) transplanted to pesticide contaminated bays form the Ebro Delta (NE, Spain).
Responses of B-esterase enzymes in oysters (Crassostrea gigas) transplanted to pesticide contaminated bays form the Ebro Delta (NE, Spain).
Marine pollution bulletin (2012-11-22)
Victoria Ochoa, Carmen Riva, Melissa Faria, Carlos Barata
ZUSAMMENFASSUNG
Marine bivalves such as oysters are widely used as bioindicators to monitor marine coastal pollution. This study aimed to use B-esterase activity responses in oysters (Crassostrea gigas) cultured in Ebro Delta bays to monitor environmental effects of pesticides. The B esterases investigated were acetylcholinesterase, propionylcholinesterase, and carboxylesterase and their activities were measured in adductor muscle and gills from oysters transplanted in Ebro Delta bays where the are traditionally grown. Enzyme activities were related with physico-chemical parameters and pesticide levels measured in water. Cholinesterase activities measured in gills were unaffected across sites and periods. Conversely, carboxylesterase activities in oyster gills varied across periods and sites and were negatively correlated with residue levels of organophoshporous and carbamate pesticides in water. Therefore, inhibition of carboxylesterase activities can be considered a good indicator of exposure to anti-cholinergic pesticides in oysters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Esterase aus Schweineleber, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Butyrylcholinesterase aus Pferdeserum, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Esterase aus Schweineleber, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Butyrylcholinesterase aus Pferdeserum, lyophilized powder, ≥900 units/mg protein
Sigma-Aldrich
Butyrylcholinesterase aus Humanserum, vial of ≥4 units
Sigma-Aldrich
Esterase aus Schweineleber, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Butyrylcholinesterase aus Pferdeserum, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Esterase aus Kaninchenleber, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esteraseisoenzym-1 aus Schweineleber, rekombinant, recombinant, expressed in E. coli, ≥30.0 U/g